missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzyl alcohol, 98%

CAS: 349-95-1 | C8H7F3O | 176.138 g/mol

$45.73 - $434.26

Chemical Identifiers

CAS 349-95-1
Molecular Formula C8H7F3O
Molecular Weight (g/mol) 176.138
MDL Number MFCD00004661
InChI Key MOOUWXDQAUXZRG-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzyl alcohol, 4-trifluoromethyl phenyl methanol, p-trifluoromethylbenzyl alcohol, 4-trifluoromethyl benzene methanol, 4-trifluoromethyl benzylic alcohol, benzenemethanol, 4-trifluoromethyl, 4-trifluoromethylbenzyl alcohol, ccris 5117, 4-trifluoromethyl benzylalcohol
PubChem CID 67684
IUPAC Name [4-(trifluoromethyl)phenyl]methanol
SMILES C1=CC(=CC=C1CO)C(F)(F)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1050806
View Documents
Thermo Scientific Chemicals
A1050806
5 g
Each for $45.73
Only null left
 
AAA1050814
View Documents
Thermo Scientific Chemicals
A1050814
25 g
Each for $153.05
Only null left
 
AAA1050822
View Documents
Thermo Scientific Chemicals
A1050822
100 g
Each for $434.26
Only null left
 
Description

Description

4-(Trifluoromethyl)benzyl alcohol, is used as a pharmaceutical intermediate, it is also used to predict the NMR spectrum. 4-(Trifluoromethyl)benzyl alcohol is employed as a reagent in, for example, kinetic studies of phosphonoformate prodrugs and aquachromium(IV).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Trifluoromethyl)benzyl alcohol, is used as a pharmaceutical intermediate, it is also used to predict the NMR spectrum. 4-(Trifluoromethyl)benzyl alcohol is employed as a reagent in, for example, kinetic studies of phosphonoformate prodrugs and aquachromium(IV).

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Keep away from oxidizing agents,reducing agents, acid chlorides, acid anhydrides.
Specifications

Chemical Identifiers

349-95-1
176.138
MOOUWXDQAUXZRG-UHFFFAOYSA-N
67684
C1=CC(=CC=C1CO)C(F)(F)F
C8H7F3O
MFCD00004661
4-trifluoromethyl benzyl alcohol, 4-trifluoromethyl phenyl methanol, p-trifluoromethylbenzyl alcohol, 4-trifluoromethyl benzene methanol, 4-trifluoromethyl benzylic alcohol, benzenemethanol, 4-trifluoromethyl, 4-trifluoromethylbenzyl alcohol, ccris 5117, 4-trifluoromethyl benzylalcohol
[4-(trifluoromethyl)phenyl]methanol

Specifications

349-95-1
1.288
100°C (212°F)
1.459
5 g
4-trifluoromethyl benzyl alcohol, 4-trifluoromethyl phenyl methanol, p-trifluoromethylbenzyl alcohol, 4-trifluoromethyl benzene methanol, 4-trifluoromethyl benzylic alcohol, benzenemethanol, 4-trifluoromethyl, 4-trifluoromethylbenzyl alcohol, ccris 5117, 4-trifluoromethyl benzylalcohol
MOOUWXDQAUXZRG-UHFFFAOYSA-N
[4-(trifluoromethyl)phenyl]methanol
67684
98%
18°C to 20°C
78°C to 80°C (4 mmHg)
C8H7F3O
MFCD00004661
1450174
Soluble in water.
C1=CC(=CC=C1CO)C(F)(F)F
176.138
176.14
4- (Trifluoromethyl)benzyl alcohol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 206-494-6

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only