Learn More
4-(Trifluoromethyl)benzyl alcohol, 98%
CAS: 349-95-1 | C8H7F3O | 176.138 g/mol
Supplier: Thermo Scientific Chemicals A1050814
Description
4-(Trifluoromethyl)benzyl alcohol, is used as a pharmaceutical intermediate, it is also used to predict the NMR spectrum. 4-(Trifluoromethyl)benzyl alcohol is employed as a reagent in, for example, kinetic studies of phosphonoformate prodrugs and aquachromium(IV).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-(Trifluoromethyl)benzyl alcohol, is used as a pharmaceutical intermediate, it is also used to predict the NMR spectrum. 4-(Trifluoromethyl)benzyl alcohol is employed as a reagent in, for example, kinetic studies of phosphonoformate prodrugs and aquachromium(IV).
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Keep away from oxidizing agents,reducing agents, acid chlorides, acid anhydrides.
Specifications
| 4- (Trifluoromethyl)benzyl alcohol | |
| 18°C to 20°C | |
| 78°C to 80°C (4 mmHg) | |
| C8H7F3O | |
| MFCD00004661 | |
| 1450174 | |
| Soluble in water. | |
| C1=CC(=CC=C1CO)C(F)(F)F | |
| 176.138 | |
| 176.14 |
| 349-95-1 | |
| 1.288 | |
| 100°C (212°F) | |
| 1.459 | |
| 25 g | |
| 4-trifluoromethyl benzyl alcohol, 4-trifluoromethyl phenyl methanol, p-trifluoromethylbenzyl alcohol, 4-trifluoromethyl benzene methanol, 4-trifluoromethyl benzylic alcohol, benzenemethanol, 4-trifluoromethyl, 4-trifluoromethylbenzyl alcohol, ccris 5117, 4-trifluoromethyl benzylalcohol | |
| MOOUWXDQAUXZRG-UHFFFAOYSA-N | |
| [4-(trifluoromethyl)phenyl]methanol | |
| 67684 | |
| 98% |
Chemical Identifiers
| 349-95-1 | |
| 176.138 | |
| MOOUWXDQAUXZRG-UHFFFAOYSA-N | |
| 67684 | |
| C1=CC(=CC=C1CO)C(F)(F)F |
| C8H7F3O | |
| MFCD00004661 | |
| 4-trifluoromethyl benzyl alcohol, 4-trifluoromethyl phenyl methanol, p-trifluoromethylbenzyl alcohol, 4-trifluoromethyl benzene methanol, 4-trifluoromethyl benzylic alcohol, benzenemethanol, 4-trifluoromethyl, 4-trifluoromethylbenzyl alcohol, ccris 5117, 4-trifluoromethyl benzylalcohol | |
| [4-(trifluoromethyl)phenyl]methanol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 206-494-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only