accessibility menu, dialog, popup

UserName

CAS RN 349-95-1

IUPAC Name: [4-(trifluoromethyl)phenyl]methanol
Synonyms: [4-(trifluoromethyl)phenyl]methanol
Molecular Weight (g/mol): 176.14
Molecular Formula: C8H7F3O
InChi Key: MOOUWXDQAUXZRG-UHFFFAOYSA-N
SMILES: OCC1=CC=C(C=C1)C(F)(F)F
Boiling Point 
  • (3)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (1)
Percent Purity 
  • (2)
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (3)
  • (1)
  • (2)
  • (5)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)