missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzoyl chloride, 97%

Catalog No. AAA1417614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1417614 25 g
AAA1417606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1417614 Supplier Thermo Scientific Chemicals Supplier No. A1417614

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 329-15-7 | C8H4ClF3O | 208.56 g/mol

4-(Trifluoromethyl)benzoyl chloride is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Trifluoromethyl)benzoyl chloride is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store under dry inert gas. Protect from humidity and water. Incompatible with water, bases, alcohol and oxidizing agents.

Chemical Identifiers

CAS 329-15-7
Molecular Formula C8H4ClF3O
Molecular Weight (g/mol) 208.56
MDL Number MFCD00000694
InChI Key OXZYBOLWRXENKT-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzoyl chloride, p-trifluoromethyl benzoyl chloride, p-trifluoromethylbenzoyl chloride, benzoyl chloride, 4-trifluoromethyl, alpha,alpha,alpha-trifluoro-p-toluoyl chloride, 4-trifluoromethyl benzoylchloride, p-trifluoromethyl benzoic acid chloride, ptf-boc, 4-trifluoromethylbenzoylchloride, 4-trifluoromethyl-benzoyl chloride
PubChem CID 67607
IUPAC Name 4-(trifluoromethyl)benzoyl chloride
SMILES FC(F)(F)C1=CC=C(C=C1)C(Cl)=O

Specifications

Melting Point -3°C
Density 1.404
Boiling Point 188°C to 190°C
Flash Point 78°C (172°F)
Odor Pungent
Refractive Index 1.477
Quantity 25 g
UN Number UN3265
Beilstein 391282
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 208.57
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethyl)benzoyl chloride
Show More Show Less
Hazard Category H227-H314
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 206-342-9
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only