missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzenesulfonyl chloride, 98%

Catalog No. AAA1290714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1290714 25 g
AAA1290706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1290714 Supplier Thermo Scientific Chemicals Supplier No. A1290714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2991-42-6 | C7H4ClF3O2S | 244.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2991-42-6
Molecular Formula C7H4ClF3O2S
Molecular Weight (g/mol) 244.61
MDL Number MFCD00042422
InChI Key OZDCZHDOIBUGAJ-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzenesulfonyl chloride, 4-trifluoromethyl benzene-1-sulfonyl chloride, 4-trifluoromethyl benzene sulfonyl chloride, p-trifluoromethylbenzenesulfonyl chloride, 4-trifluoromethyl benzenesulfonylchloride, 4-trifluoromethyl-benzenesulfonyl chloride, 4-trifluoromethyl phenylsulfonyl chloride, p-trifluoromethyl benzenesulfonyl chloride, 4-trifluoromethyl benzenesulphonyl chloride, benzenesulfonyl chloride, 4-trifluoromethyl
PubChem CID 2777399
IUPAC Name 4-(trifluoromethyl)benzenesulfonyl chloride
SMILES FC(F)(F)C1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

Melting Point 28°C to 34°C
Boiling Point 118°C to 120°C (18 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
UN Number UN3261
Beilstein 2113604
Sensitivity Moisture sensitive
Formula Weight 244.62
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethyl)benzenesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only