missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzenesulfonamide, 97%

Catalog No. p-7028107
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2524303 1 g
AAB2524314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2524303 Supplier Thermo Scientific Chemicals Supplier No. B2524303

CAS: 830-43-3 | C7H6F3NO2S | 225.185 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 830-43-3
Molecular Formula C7H6F3NO2S
Molecular Weight (g/mol) 225.185
MDL Number MFCD00159251
InChI Key TVHXQQJDMHKGGK-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzenesulfonamide, 4-trifluoromethyl benzene-1-sulfonamide, benzenesulfonamide, p-trifluoromethyl, 4-trifluoromethylbenzenesulphonamide, p-trifluoromethyl benzenesulfonamide, 4-trifluoromethyl benzenesulphonamide, benzenesulfonamide, 4-trifluoromethyl, p-trifluoromethylbenzenesulfonamide, alpha,alpha,alpha-trifluoro-p-toluenesulphonamide, pubchem11767
PubChem CID 70018
IUPAC Name 4-(trifluoromethyl)benzenesulfonamide
SMILES C1=CC(=CC=C1C(F)(F)F)S(=O)(=O)N

Specifications

Melting Point 178°C to 182°C
Quantity 1 g
Beilstein 2695323
Formula Weight 225.19
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethyl)benzenesulfonamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 212-596-1
RTECSNumber DB3200000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only