missing translation for 'onlineSavingsMsg'
Learn More

4-Trifluoromethyl-2,3,5,6-tetrafluorobromobenzene 99.0+%, TCI America™

Catalog No. T154110G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T154110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T154110G Supplier TCI America Supplier No. T154110G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17823-46-0
Molecular Formula C7BrF7
Molecular Weight (g/mol) 296.97
MDL Number MFCD00075240
InChI Key DONVNRFILRLHJB-UHFFFAOYSA-N
Synonym 1-bromo-2,3,5,6-tetrafluoro-4-trifluoromethyl benzene, 4-bromo-2,3,5,6-tetrafluorobenzotrifluoride, p-bromoheptafluorotoluene, 4-trifluoromethyl-2,3,5,6-tetrafluorobromobenzene, acmc-209ee7, 4-bromoheptafluorotoluene, 4-bromo-alpha,alpha,alpha,2,3,5,6-heptafluorotoluene, 1-trifluoromethyl-2,3,5,6-tetrafluoro-4-bromobenzene
PubChem CID 626637
IUPAC Name 1-bromo-2,3,5,6-tetrafluoro-4-(trifluoromethyl)benzene
SMILES C1(=C(C(=C(C(=C1F)F)Br)F)F)C(F)(F)F

Specifications

Color Colorless
Boiling Point 153°C
Quantity 10 g
Formula Weight 296.97
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Trifluoromethyl-2,3,5,6-tetrafluorobromobenzene