missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. T17735G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T17735G Supplier TCI America Supplier No. T17735G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 139301-27-2
Molecular Formula C7H6BF3O3
Molecular Weight (g/mol) 205.93
MDL Number MFCD01074648
InChI Key HUOFUOCSQCYFPW-UHFFFAOYSA-N
PubChem CID 2734386
IUPAC Name [4-(trifluoromethoxy)phenyl]boronic acid
SMILES OB(O)C1=CC=C(OC(F)(F)F)C=C1

Specifications

Melting Point 127°C
Color White-Yellow
Quantity 5 g
Synonym 4-trifluoromethoxy phenylboronic acid, 4-trifluoromethoxyphenylboronic acid, 4-trifluoromethoxy benzeneboronic acid, 4-trifluoromethoxy phenyl boronic acid, 4-trifluoromethoxy phenyl boranediol, p-trifluoromethoxy phenylboronic acid, 4-trifluoromethoxy-phenyl boronic acid, boronic acid, 4-trifluoromethoxy phenyl, 4-trifluoromethoxy phenylboronicacid
Formula Weight 205.93
Physical Form Crystalline Powder
Chemical Name or Material 4-(Trifluoromethoxy)phenylboronic Acid (contains varying amounts of Anhydride)