missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)benzoyl Chloride 98.0+%, TCI America™

Catalog No. T23415G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23415G 5 g
T23411G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T23415G Supplier TCI America Supplier No. T23415G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 36823-88-8
Molecular Formula C8H4ClF3O2
Molecular Weight (g/mol) 224.563
MDL Number MFCD00052329
InChI Key ZXKKOFJYPRJFIE-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy benzoyl chloride, p-trifluoromethoxybenzoyl chloride, benzoyl chloride, 4-trifluoromethoxy, 4-trifluoromethoxy benzene-1-carbonyl chloride, 4-trifluoromethoxy benzoylchloride, 4-trifluoromethoxy-benzoyl chloride, p-trifluoromethoxy benzoyl chloride, acmc-209ip1, ksc495i1f, p-trifluoromethoxybenzoylchloride
PubChem CID 142110
IUPAC Name 4-(trifluoromethoxy)benzoyl chloride
SMILES C1=CC(=CC=C1C(=O)Cl)OC(F)(F)F

Specifications

Color Colorless
Boiling Point 56°C
Quantity 5 g
UN Number 3265
Formula Weight 224.56
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-(Trifluoromethoxy)benzoyl Chloride