missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)benzonitrile 98.0+%, TCI America™

Catalog No. T22925G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T22925G 5 g
T22921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T22925G Supplier TCI America Supplier No. T22925G
Only null left

Chemical Identifiers

CAS 332-25-2
Molecular Formula C8H4F3NO
Molecular Weight (g/mol) 187.121
MDL Number MFCD00039474
InChI Key XWHIXOMWXCHJPP-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy benzonitrile, benzonitrile, 4-trifluoromethoxy, p-trifluoromethoxybenzonitrile, p-cyanotrifluoromethoxybenzene, p-trifluoromethoxy benzonitrile, 1-cyano-4-trifluoromethoxy benzene, ncr doxfff, 4-trifluoromethoxy benzenecarbonitrile, pubchem4826, acmc-1coco
PubChem CID 67621
IUPAC Name 4-(trifluoromethoxy)benzonitrile
SMILES C1=CC(=CC=C1C#N)OC(F)(F)F

Specifications

Color Yellow
Boiling Point 193°C
Quantity 5 g
UN Number 3276
Formula Weight 187.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-(Trifluoromethoxy)benzonitrile