missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoroacetyl)benzoic acid, 97+%

Catalog No. AAL10242MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL10242MD 250 mg
AAL1024203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL10242MD Supplier Thermo Scientific Chemicals Supplier No. L10242MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 58808-59-6 | C9H5F3O3 | 218.131 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 58808-59-6
Molecular Formula C9H5F3O3
Molecular Weight (g/mol) 218.131
MDL Number MFCD00052340
InChI Key WLTZCRCZDLLXQP-UHFFFAOYSA-N
Synonym 4-trifluoroacetyl benzoic acid, 4-2,2,2-trifluoroacetyl benzoic acid, benzoic acid, 4-trifluoroacetyl, 4-2,2,2-trifluoro-acetyl-benzoic acid, benzoic acid, 4-trifluoroacetyl-9ci, 4-carboxy-a,a,a-trifluoroacetophenone, pubchem13997, acmc-1atst, 4-trifluoroacetylbenzoic acid, labotest-bb lt00454127
PubChem CID 143579
IUPAC Name 4-(2,2,2-trifluoroacetyl)benzoic acid
SMILES C1=CC(=CC=C1C(=O)C(F)(F)F)C(=O)O

Specifications

Melting Point 177°C to 179°C
Quantity 250 mg
Beilstein 2728778
Sensitivity Hygroscopic
Formula Weight 218.13
Percent Purity ≥97%
Chemical Name or Material 4-(Trifluoroacetyl)benzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only