missing translation for 'onlineSavingsMsg'
Learn More

4-(trans-4-Vinylcyclohexyl)benzonitrile 98.0+%, TCI America™

Catalog No. V01355G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V01355G 5 g
V01351G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V01355G Supplier TCI America Supplier No. V01355G

Chemical Identifiers

CAS 96184-42-8
Molecular Formula C15H17N
Molecular Weight (g/mol) 211.308
MDL Number MFCD11053490
InChI Key JHXJAWXZLWDDED-UHFFFAOYSA-N
Synonym 4-trans-4-vinylcyclohexyl benzonitrile, trans-4'-4-vinylcyclohexyl benzonitrile, trans-4-4-vinyl-cyclohexyl-benzonitrile, 4-4-ethenylcyclohexyl benzonitrile, 4-4-vinylcyclohexyl benzonitrile, jhxjawxzlwdded-mqmhxkeqsa-n, p-trans-4-vinylcyclohexyl benzonitrile, trans-4-4-vinylcyclohexyl benzonitrile, p-trans-4-vinylcyclohexyl-benzonitrile, 4-trans-4-ethenylcyclohexyl benzonitrile
PubChem CID 18724353
IUPAC Name 4-(4-ethenylcyclohexyl)benzonitrile
SMILES C=CC1CCC(CC1)C2=CC=C(C=C2)C#N

Specifications

Melting Point 58°C
Color White
Quantity 5 g
Formula Weight 211.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(trans-4-Vinylcyclohexyl)benzonitrile