missing translation for 'onlineSavingsMsg'
Learn More

4-(thien-2-ylmethyl)aniline, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC70614DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC70614DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

129136-65-8
189.276
4-thien-2-ylmethyl aniline, 4-thiophen-2-ylmethyl aniline, 4-thiophen-2-yl methyl aniline, 4-2-thienylmethyl phenylamine
4-(thiophen-2-ylmethyl)aniline
C11H11NS
PIJZLXNJWCKMSA-UHFFFAOYSA-N
20063869
C1=CSC(=C1)CC2=CC=C(C=C2)N

Specifications

4-(Thien-2-ylmethyl)aniline
95%
C11H11NS
4-thien-2-ylmethyl aniline, 4-thiophen-2-ylmethyl aniline, 4-thiophen-2-yl methyl aniline, 4-2-thienylmethyl phenylamine
C1=CSC(=C1)CC2=CC=C(C=C2)N
189.276
189.28
129136-65-8
Amber Glass Bottle
1g
PIJZLXNJWCKMSA-UHFFFAOYSA-N
4-(thiophen-2-ylmethyl)aniline
20063869
95%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS
Documents

Documents