missing translation for 'onlineSavingsMsg'
Learn More

4-(thien-2-ylmethyl)aniline, Thermo Scientific™

Catalog No. CC70614CB
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC70614CB 250mg Amber Glass Bottle
CC70614DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC70614CB Supplier Thermo Scientific Chemicals Supplier No. CC70614CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 129136-65-8
Molecular Formula C11H11NS
Molecular Weight (g/mol) 189.276
InChI Key PIJZLXNJWCKMSA-UHFFFAOYSA-N
Synonym 4-thien-2-ylmethyl aniline, 4-thiophen-2-ylmethyl aniline, 4-thiophen-2-yl methyl aniline, 4-2-thienylmethyl phenylamine
PubChem CID 20063869
IUPAC Name 4-(thiophen-2-ylmethyl)aniline
SMILES C1=CSC(=C1)CC2=CC=C(C=C2)N

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 189.28
Percent Purity 95%
Chemical Name or Material 4-(Thien-2-ylmethyl)aniline
Hazard Category GHS Signal Word: Warning