missing translation for 'onlineSavingsMsg'
Learn More

4-(Tetrahydropyran-4-yloxy)benzonitrile, 97%, Thermo Scientific™

Numéro de catalogue. p-3830103
Modifier l'affichage
Click to view available options
Quantité:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CC52416DA 1g
CC52416DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. CC52416DA Fournisseur Thermo Scientific Chemicals Code fournisseur CC52416DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 884507-34-0
Formule moléculaire C12H13NO2
Poids moléculaire (g/mol) 203.241
Numéro MDL MFCD09025875
Clé InChI ITXQSYHCKPGZLV-UHFFFAOYSA-N
Synonyme 4-tetrahydropyran-4-yloxy benzonitrile, 4-oxan-4-yloxy benzonitrile, 4-tetrahydro-2h-pyran-4-yloxy benzonitrile, 4-oxan-4-yl oxy benzonitrile, 4-tetrahydro-2h-pyran-4-yl oxy benzonitrile, benzonitrile,4-tetrahydro-2h-pyran-4-yl oxy, 4-2h-3,4,5,6-tetrahydropyran-4-yloxy benzenecarbonitrile
PubChem CID 24229527
Nom de l’IUPAC 4-(oxan-4-yloxy)benzonitrile
SOURIRES C1COCCC1OC2=CC=C(C=C2)C#N

Spécifications

Nom ou substance chimique 4-(Tetrahydropyran-4-yloxy)benzonitrile
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber glass bottle
Quantité 1g
Poids de formule 203.24
Pourcentage de pureté 97%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.