missing translation for 'onlineSavingsMsg'
Learn More

4-(Tetrahydropyran-4-yloxy)benzonitrile, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC52416DE

Quantity 5g

 View more versions of this product

Catalog No. CC52416DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

884507-34-0
203.241
ITXQSYHCKPGZLV-UHFFFAOYSA-N
24229527
C1COCCC1OC2=CC=C(C=C2)C#N
C12H13NO2
MFCD09025875
4-tetrahydropyran-4-yloxy benzonitrile, 4-oxan-4-yloxy benzonitrile, 4-tetrahydro-2h-pyran-4-yloxy benzonitrile, 4-oxan-4-yl oxy benzonitrile, 4-tetrahydro-2h-pyran-4-yl oxy benzonitrile, benzonitrile,4-tetrahydro-2h-pyran-4-yl oxy, 4-2h-3,4,5,6-tetrahydropyran-4-yloxy benzenecarbonitrile
4-(oxan-4-yloxy)benzonitrile

Specifications

4-(Tetrahydropyran-4-yloxy)benzonitrile
96.5
97%
5g
4-tetrahydropyran-4-yloxy benzonitrile, 4-oxan-4-yloxy benzonitrile, 4-tetrahydro-2h-pyran-4-yloxy benzonitrile, 4-oxan-4-yl oxy benzonitrile, 4-tetrahydro-2h-pyran-4-yl oxy benzonitrile, benzonitrile,4-tetrahydro-2h-pyran-4-yl oxy, 4-2h-3,4,5,6-tetrahydropyran-4-yloxy benzenecarbonitrile
C1COCCC1OC2=CC=C(C=C2)C#N
203.241
203.24
884507-34-0
100.0
C12H13NO2
MFCD09025875
ITXQSYHCKPGZLV-UHFFFAOYSA-N
4-(oxan-4-yloxy)benzonitrile
24229527
97%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents