missing translation for 'onlineSavingsMsg'
Learn More

4-(Tetrahydropyran-4-yloxy)aniline, 97%, Thermo Scientific™

Catalog No. CC52414CB
Change view
Click to view available options
Amber Glass Bottle; 250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
CC52414CB Amber Glass Bottle; 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC52414CB Supplier Thermo Scientific Chemicals Supplier No. CC52414CB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

missing translation for 'validMainframeMsg'

Specifications

Nom ou substance chimique 4-(Tetrahydropyran-4-yloxy)aniline
CAS 917483-71-7
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber Glass Bottle
Formule moléculaire C11H15NO2
Quantité 250mg
Numéro MDL MFCD09064968
Synonyme 4-tetrahydropyran-4-yloxy aniline, 4-oxan-4-yloxy aniline, 4-tetrahydro-2h-pyran-4-yl oxy aniline, 4-oxan-4-yl oxy aniline, 4-tetrahydropyran-4-yloxy phenylamine, 4-tetrahydro-2h-pyran-4-yloxy aniline, 4-tetrahydro-pyran-4-yloxy-phenylamine, 4-2h-3,4,5,6-tetrahydropyran-4-yloxy phenylamine
Clé InChI CCIFXNITZHWCRR-UHFFFAOYSA-N
SOURIRES C1COCCC1OC2=CC=C(C=C2)N
Nom de l’IUPAC 4-(oxan-4-yloxy)aniline
Poids moléculaire (g/mol) 193.246
PubChem CID 24229526
Poids de formule 193.24
Pourcentage de pureté 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.