missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butyltoluene 95.0+%, TCI America™

Catalog No. B0744500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0744500ML 500 mL
B074425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0744500ML Supplier TCI America Supplier No. B0744500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 98-51-1
Molecular Formula C11H16
Molecular Weight (g/mol) 148.249
MDL Number MFCD00008837
InChI Key QCWXDVFBZVHKLV-UHFFFAOYSA-N
Synonym 4-tert-butyltoluene, p-tert-butyltoluene, 1-tert-butyl-4-methylbenzene, p-t-butyltoluene, benzene, 1-1,1-dimethylethyl-4-methyl, 4-t-butyltoluene, p-methyl-tert-butylbenzene, toluene, p-tert-butyl, 1-methyl-4-tert-butylbenzene, 4-methyl-tert-butylbenzene
PubChem CID 7390
IUPAC Name 1-tert-butyl-4-methylbenzene
SMILES CC1=CC=C(C=C1)C(C)(C)C

Specifications

Color Yellow
Boiling Point 193°C
Quantity 500 mL
UN Number 2667
Formula Weight 148.25
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-tert-Butyltoluene
EINECSNumber (3)-0015
RTECSNumber XS8400000
TSCA Yes