missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylstyrene (stabilized with TBC) 90.0+%, TCI America™

Catalog No. B1877500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1877500ML 500 mL
B187725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1877500ML Supplier TCI America Supplier No. B1877500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1746-23-2
Molecular Formula C12H16
Molecular Weight (g/mol) 160.26
MDL Number MFCD00065126
InChI Key QEDJMOONZLUIMC-UHFFFAOYSA-N
Synonym 4-tert-butylstyrene, p-tert-butylstyrene, p-t-butylstyrene, benzene, 1-1,1-dimethylethyl-4-ethenyl, styrene, p-tert-butyl, unii-9pct9cbw0v, 1-1,1-dimethylethyl-4-ethenylbenzene, 9pct9cbw0v, 1-tert-butyl-4-vinylbenzene, 4-t-butyl styrene
PubChem CID 15627
IUPAC Name 1-tert-butyl-4-ethenylbenzene
SMILES CC(C)(C)C1=CC=C(C=C1)C=C

Specifications

Color Yellow
Quantity 500 mL
Formula Weight 160.26
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-tert-Butylstyrene (stabilized with TBC)
TSCA Yes
Recommended Storage Freezer