missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylpyridine 96.0+%, TCI America™

Catalog No. B03885G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B03885G 5 g
B038825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B03885G Supplier TCI America Supplier No. B03885G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3978-81-2
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00006435
InChI Key YSHMQTRICHYLGF-UHFFFAOYSA-N
Synonym 4-tert-butyl pyridine, pyridine, 4-1,1-dimethylethyl, 4-t-butylpyridine, pyridine, 4-tert-butyl, p-tert-butyl pyridine, unii-il8v02163i, 4-tert-butyl-pyridine, 4-t-butyl pyridine, 4-tert.butylpyridine
PubChem CID 19878
IUPAC Name 4-tert-butylpyridine
SMILES CC(C)(C)C1=CC=NC=C1

Specifications

Melting Point -40°C
Color Yellow
Boiling Point 197°C
Quantity 5 g
Formula Weight 135.21
Percent Purity ≥96.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-tert-Butylpyridine