missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylphenoxyacetic acid, 98%

Catalog No. AAA1701722
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1701722 100 g
AAA1701706 5 g
AAA1701714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1701722 Supplier Thermo Scientific Chemicals Supplier No. A1701722
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1798-04-5 | C12H16O3 | 208.257 g/mol

Applied as a reagent for the TAC protective group in nucleosides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Applied as a reagent for the TAC protective group in nucleosides.

Solubility
Sparingly Soluble (0.56 g/L) (25°C), Calc.

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.

Chemical Identifiers

CAS 1798-04-5
Molecular Formula C12H16O3
Molecular Weight (g/mol) 208.257
MDL Number MFCD00021758
InChI Key FBIGAJNVRFKBJL-UHFFFAOYSA-N
Synonym 4-tert-butylphenoxyacetic acid, 4-tert-butylphenoxy acetic acid, 4-tert-butyl-phenoxy-acetic acid, 2-4-tert-butylphenoxy acetic acid, p-tert-butylphenoxy acetic acid, p-t-butylphenoxyacetic acid, 2-4-tert-butyl phenoxy acetic acid, acetic acid, 4-1,1-dimethylethyl phenoxy, acetic acid, 2-4-1,1-dimethylethyl phenoxy
PubChem CID 15718
IUPAC Name 2-(4-tert-butylphenoxy)acetic acid
SMILES CC(C)(C)C1=CC=C(C=C1)OCC(=O)O

Specifications

Quantity 100 g
Solubility Information Sparingly Soluble (0.56g/L) (25°C),Calc.
Formula Weight 208.26
Percent Purity 98%
Chemical Name or Material 4-tert-Butylphenoxyacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only