missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylphenol Zone Refined (number of passes:19) 98.0+%, TCI America™

Catalog No. B0735ZONE
Change view
Click to view available options
Quantity:
ZONE
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0735ZONE ZONE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B0735ZONE Supplier TCI America Supplier No. B07351SAMPLE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 98-54-4
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00002367
InChI Key QHPQWRBYOIRBIT-UHFFFAOYSA-N
Synonym 4-tert-butyl phenol, butylphen, p-tert-butylphenol, ptbp, 4-1,1-dimethylethyl phenol, 4-t-butylphenol, p-t-butyl phenol, phenol, 4-1,1-dimethylethyl, p-t-butylphenol, p-terc.butylfenol
PubChem CID 7393
ChEBI CHEBI:34444
IUPAC Name 4-tert-butylphenol
SMILES CC(C)(C)C1=CC=C(C=C1)O

Specifications

Melting Point 101°C
Color White
Boiling Point 238°C
Quantity ZONE
UN Number 2430
Formula Weight 150.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-tert-Butylphenol Zone Refined (number of passes:19)
EINECSNumber (3)-0503
RTECSNumber SJ8925000
TSCA Yes