missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylcyclohexanol (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. B038325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B038325G 25 g
B0383500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B038325G Supplier TCI America Supplier No. B038325G

Chemical Identifiers

CAS 98-52-2
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.27
MDL Number MFCD00001473,MFCD00064952,MFCD00070476
InChI Key CCOQPGVQAWPUPE-UHFFFAOYSA-N
Synonym 4-tert-butylcyclohexanol, cis-4-tert-butylcyclohexanol, trans-4-tert-butylcyclohexanol, 4-tert-butyl cyclohexanol, cis-4-tert-butyl cyclohexanol, padaryl, p-tert-butylcyclohexanol, cyclohexanol, 4-1,1-dimethylethyl, cyclohexanol, 4-tert-butyl, 4-t-butylcyclohexanol
PubChem CID 7391
IUPAC Name 4-tert-butylcyclohexan-1-ol
SMILES CC(C)(C)C1CCC(O)CC1

Specifications

Melting Point 68°C
Color White
Quantity 25 g
Formula Weight 156.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-tert-Butylcyclohexanol (cis- and trans- mixture)
EINECSNumber (3)-2301
RTECSNumber GV8750000
TSCA Yes