missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylcyclohexanecarboxylic Acid (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. B12855G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B12855G 5 g
B128525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B12855G Supplier TCI America Supplier No. B12855G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5451-55-8
Molecular Formula C11H20O2
Molecular Weight (g/mol) 184.279
MDL Number MFCD00042622
InChI Key QVQKEGYITJBHRQ-UHFFFAOYSA-N
Synonym trans-4-tert-butylcyclohexanecarboxylic acid, 4-tert-butylcyclohexanecarboxylic acid, cis-4-tert-butylcyclohexanecarboxylic acid, trans-4-tert-butyl cyclohexanecarboxylic acid, 4-tert-butyl cyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 4-1,1-dimethylethyl-, cis, cyclohexanecarboxylic acid, 4-1,1-dimethylethyl, cis-4-tert-butylcyclohexane carboxylic acid, cyclohexanecarboxylic acid, 4-1,1-dimethylethyl-, trans, qvqkegyitjbhrq-dtorhvgosa-n
PubChem CID 136759
IUPAC Name 4-tert-butylcyclohexane-1-carboxylic acid
SMILES CC(C)(C)C1CCC(CC1)C(=O)O

Specifications

Color White
Quantity 5 g
Formula Weight 184.28
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-tert-Butylcyclohexanecarboxylic Acid (cis- and trans- mixture)
EINECSNumber (3)-3723
TSCA No