missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzohydrazide 98.0+%, TCI America™

Catalog No. B28865G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28865G 5 g
B288625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28865G Supplier TCI America Supplier No. B28865G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 43100-38-5
Molecular Formula C11H16N2O
Molecular Weight (g/mol) 192.262
MDL Number MFCD00014763
InChI Key XYUFQWDLRLHUPB-UHFFFAOYSA-N
Synonym 4-tert-butylbenzhydrazide, 4-tert-butyl benzohydrazide, benzoic acid, 4-1,1-dimethylethyl-, hydrazide, p-tert-butylbenzohydrazide, 4-tert-butylbenzhyrazide, 4-tert-butylbenzoic acid hydrazide, 4-tert-butylbenzoic hydrazide, 4-tert-butyl benzenecarbohydrazide, 4-tert-butyl benzene-1-carbohydrazide, 4-t-butylbenzhydrazide
PubChem CID 123513
IUPAC Name 4-tert-butylbenzohydrazide
SMILES CC(C)(C)C1=CC=C(C=C1)C(=O)NN

Specifications

Melting Point 130°C
Color White
Quantity 5 g
Formula Weight 192.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-tert-Butylbenzohydrazide
EINECSNumber (3)-4326
TSCA Yes