missing translation for 'onlineSavingsMsg'
Learn More

4-(tert-Butyl)-1,3-thiazol-2-amine, 97%, Thermo Scientific™

Numéro de catalogue. p-2700872
Modifier l'affichage
Click to view available options
Quantité:
1g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CD08308DA 1g
CD08308EA 10g
CD08308EB 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. CD08308DA Fournisseur Thermo Scientific Chemicals Code fournisseur CD08308DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 74370-93-7
Formule moléculaire C7H12N2S
Poids moléculaire (g/mol) 156.247
Numéro MDL MFCD00051644
Clé InChI CUWZBHVYLVGOAB-UHFFFAOYSA-N
Synonyme 2-amino-4-tert-butylthiazole, 4-tert-butylthiazol-2-ylamine, 4-tert-butyl thiazol-2-amine, 4-tert-butyl-thiazol-2-ylamine, 2-amino-4-t-butylthiazole, 4-tert-butyl-1,3-thiazol-2-amine, 4-tert-butylthiazol-2-amine, 4-tert-butyl-1,3-thiazol-2-ylamine, 2-thiazolamine, 4-1,1-dimethylethyl, 4-tert-butyl-1,3-thiazole-2-ylamine
PubChem CID 2734202
Nom de l’IUPAC 4-tert-butyl-1,3-thiazol-2-amine
SOURIRES CC(C)(C)C1=CSC(=N1)N

Spécifications

Nom ou substance chimique 4-(tert-Butyl)-1, 3-thiazol-2-amine
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber glass bottle
Quantité 1g
Poids de formule 156.25
Pourcentage de pureté 97%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.