missing translation for 'onlineSavingsMsg'
Learn More

4-(tert-Butoxycarbonylamino)benzoic Acid 98.0+%, TCI America™

Catalog No. B38985G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38985G 5 g
B389825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38985G Supplier TCI America Supplier No. B38985G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 66493-39-8
Molecular Formula C12H15NO4
Molecular Weight (g/mol) 237.255
MDL Number MFCD00037428
InChI Key ZJDBQMWMDZEONW-UHFFFAOYSA-N
Synonym boc-4-abz-oh, 4-boc-amino benzoic acid, 4-tert-butoxycarbonyl amino benzoic acid, 4-tert-butoxycarbonylamino-benzoic acid, n-boc-4-aminobenzoic acid, 4-tert-butoxycarbonylaminobenzoic acid, 4-tert-butoxycarbonylamino benzoic acid, boc-p-aminobenzoic acid, 4-boc-amino-benzoic acid
PubChem CID 2755931
IUPAC Name 4-[(2-methylpropan-2-yl)oxycarbonylamino]benzoic acid
SMILES CC(C)(C)OC(=O)NC1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 196°C
Color White
Quantity 5 g
Formula Weight 237.26
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(tert-Butoxycarbonylamino)benzoic Acid