missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Amylphenol, 99%

Catalog No. AC167330050
Change view
Click to view available options
Quantity:
5 g
100 g
250 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC167330050 5 g
AC167331000 100 g
AC167332500 250 g
AC167335000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC167330050 Supplier Thermo Scientific Chemicals Supplier No. 167330050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 80-46-6 | C11H16O | 164.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 80-46-6
Molecular Formula C11H16O
Molecular Weight (g/mol) 164.25
MDL Number MFCD00002369
InChI Key NRZWYNLTFLDQQX-UHFFFAOYSA-N
Synonym 4-tert-amylphenol, 4-tert-pentylphenol, p-tert-amylphenol, p-tert-pentylphenol, 4-t-amylphenol, 4-1,1-dimethylpropyl phenol, amilphenol, amilfenol, pentaphen, tert-amylphenol
PubChem CID 6643
ChEBI CHEBI:35096
IUPAC Name 4-(2-methylbutan-2-yl)phenol
SMILES CCC(C)(C)C1=CC=C(C=C1)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point >91.0°C
Color White to Yellow
Boiling Point 255.0°C
Flash Point 111°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C2H5C(CH3)2C6H4OH
Quantity 5 g
Beilstein 06, 548
Merck Index 15, 7255
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, ether, benzene and chloroform
Viscosity 2.9 mPa.s (100°C)
Formula Weight 164.25
Percent Purity 99%
Physical Form Briquettes or Flakes to Coarse Powder
Chemical Name or Material 4-tert-Amylphenol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several m
EINECSNumber 201-280-9
RTECSNumber SM6825000
TSCA TSCA

RUO – Research Use Only