missing translation for 'onlineSavingsMsg'
Learn More

4-Sulfobenzoic acid monopotassium salt, 95%

Catalog No. AAA1943218
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1943218 50 g
AAA1943230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1943218 Supplier Thermo Scientific Chemicals Supplier No. A1943218
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5399-63-3 | C7H5KO5S | 240.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5399-63-3
Molecular Formula C7H5KO5S
Molecular Weight (g/mol) 240.27
MDL Number MFCD00007509
InChI Key PXRJBUPXKDXDLG-UHFFFAOYSA-M
Synonym benzoic acid, 4-sulfo-, monopotassium salt, 4-sulfobenzoic acid monopotassium salt, 4-sulphobenzoic acid monopotassium salt, 4-sulfobenzoicacidmonopotassiumsalt, potassium hydrogen 4-sulphonatobenzoate, p-sulphobenzoic acid, monopotassium salt, potassium 4-sulphobenzoate, benzoic acid, 4-sulfo-, potassium salt 1:1, potassium 4-sulfobenzoate, p-potassiooxysulfonyl benzoic acid
PubChem CID 23670833
IUPAC Name potassium;4-sulfobenzoate
SMILES C1=CC(=CC=C1C(=O)[O-])S(=O)(=O)O.[K+]

Specifications

Quantity 50 g
Sensitivity Hygroscopic
Formula Weight 240.28
Percent Purity 95%
Chemical Name or Material 4-Sulfobenzoic acid monopotassium salt
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 226-435-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only