missing translation for 'onlineSavingsMsg'
Learn More

4-Ring NOGE (Novolac glycidyl ether) Mixture of Isomers, chain-like or branched, ≥90% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111003392
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111003392 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111003392 Supplier Merck Emd Millipore Supplier No. 0497650MGF
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

Synonym NOGE 4-Ring (Novolac glycidyl ether) mixture of isomers; Novolac glycidyl ether 4-Ring
Type Analytical Standard
Quantity 50 mg
Percent Purity ≥90% (HPLC)
Grade Analytical Standard