missing translation for 'onlineSavingsMsg'
Learn More

4-Pyrimidin-5-ylbenzoic acid, ≥97%, Thermo Scientific™

Numéro de catalogue. p-3176032
Change view
Click to view available options
Quantité:
250mg
1g
Emballage:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité Emballage
CC44301CB 250mg Amber Glass Bottle
CC44301DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC44301CB Supplier Thermo Scientific Chemicals Supplier No. CC44301CB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 216959-91-0
Formule moléculaire C11H8N2O2
Poids moléculaire (g/mol) 200.197
Numéro MDL MFCD05864811
Clé InChI CCBKADQVSXLSDN-UHFFFAOYSA-N
Synonyme 4-pyrimidin-5-yl benzoic acid, 4-pyrimidin-5-yl-benzoic acid, 4-5-pyrimidinyl benzoic acid, benzoic acid, 4-5-pyrimidinyl
PubChem CID 4187247
Nom de l’IUPAC 4-pyrimidin-5-ylbenzoic acid
SOURIRES C1=CC(=CC=C1C2=CN=CN=C2)C(=O)O

Specifications

Nom ou substance chimique 4-Pyrimidin-5-ylbenzoic acid
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber Glass Bottle
Quantité 250mg
Poids de formule 200.2
Pourcentage de pureté 97%
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.