missing translation for 'onlineSavingsMsg'
Learn More

4-Pyrimidin-2-ylbenzonitrile, 95%, Thermo Scientific™

Catalog No. CC56216DA
Change view
Click to view available options
Quantity:
1g
5g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC56216DA 1g Amber Glass Bottle
CC56216DE 5g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC56216DA Supplier Thermo Scientific Chemicals Supplier No. CC56216DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 78322-96-0
Molecular Formula C11H7N3
Molecular Weight (g/mol) 181.198
MDL Number MFCD09817491
InChI Key WGCKVXHOTDDNOA-UHFFFAOYSA-N
Synonym 4-pyrimidin-2-yl benzonitrile, 4-cyanophenyl pyrimidine, 2-4-cyanophenyl pyrimidine, 4-pyrimidin-2-yl-benzonitrile, 4-2-pyrimidinyl benzonitrile, benzonitrile, 4-2-pyrimidinyl, 4-pyrimidin-2-ylbenzenecarbonitrile
PubChem CID 15486310
IUPAC Name 4-pyrimidin-2-ylbenzonitrile
SMILES C1=CN=C(N=C1)C2=CC=C(C=C2)C#N

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 181.2
Percent Purity 95%
Chemical Name or Material 4-Pyrimidin-2-ylbenzonitrile
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.