missing translation for 'onlineSavingsMsg'
Learn More

4-Pyridin-4-ylbenzaldehyde, 97%, Thermo Scientific™

Catalog No. CC35404CB
Change view
Click to view available options
Amber Glass Bottle; 250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
CC35404CB Amber Glass Bottle; 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC35404CB Supplier Thermo Scientific Chemicals Supplier No. CC35404CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 99163-12-9
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C12H9NO
Quantity 250mg
Synonym 4-pyridin-4-yl-benzaldehyde, 4-pyridin-4-yl benzaldehyde, 4-4-pyridyl benzaldehyde, 4-4-pyridinyl benzaldehyde, 4-4-formylphenyl pyridine, akos bar-0607, benzaldehyde, 4-4-pyridinyl, acmc-209sbt
InChI Key MBJXDIYHLGBQOT-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1C=O)C2=CC=NC=C2
IUPAC Name 4-pyridin-4-ylbenzaldehyde
Molecular Weight (g/mol) 183.21
PubChem CID 639673
Formula Weight 183.21
Percent Purity 97%
Chemical Name or Material 4-Pyridin-4-ylbenzaldehyde
Show More Show Less