Learn More
Eisco™ 4 Piece Metalware Set
Catalog No. S23711
Change view
Click to view available options
Height (Metric):
61 cm
Catalog No. Height (Metric)
S23711 61 cm
1 options
Catalog No. S23711 Supplier Eisco™ Supplier No. MTST5
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Choose durable lab equipment that will last for years. The EISCO 4 Piece Metalware Set is a durable and economical metalware set that is a great start to any laboratory.
  • Simple metal ware set for laboratory use
  • Kit consists of: (1) Base 20 X 12.5cm, (1) Steel Rod 60cm, (1) Retort Ring 7.5cm Outer Diameter, (1) 3 Coated Prong Clamp with boss head
  • Powder coated, chemical resistant base
  • Plated steel rod for chemical resistance and years of use
  • 3 Prong clamp is able to swivel, coated tips will protect glass, and wide mouth will accommodate most common glassware up to 3.5 inches wide
Specifications
Material Powder-coated Chemical Resistant (Base)
Height (Metric) 61 cm
Includes (1) Base 20cm x 12.5cm, (1) Steel Rod 60cm, (1) Retort Ring 7.5cm Outer Diameter, (1) 3 Coated Prong Clamp with Boss Head