missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyltetrahydropyran-4-carboxaldehyde, 95%

Catalog No. AAH50816MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH50816MD 250 mg
AAH5081603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH50816MD Supplier Thermo Scientific Chemicals Supplier No. H50816MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 66109-88-4 | C12H14O2 | 190.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 66109-88-4
Molecular Formula C12H14O2
Molecular Weight (g/mol) 190.24
MDL Number MFCD08061006
InChI Key IEIVCCVSMYLGOC-UHFFFAOYSA-N
Synonym 4-phenyltetrahydro-2h-pyran-4-carboxaldehyde, 4-phenyltetrahydro-2h-pyran-4-carbaldehyde, 4-phenyltetrahydropyran-4-carbaldehyde, 4-phenyl-2h-3,4,5,6-tetrahydropyran-4-carbaldehyde, acmc-1b39w, tetrahydro-4-phenyl-2h-pyran-4-carboxaldehyde
PubChem CID 12405610
IUPAC Name 4-phenyloxane-4-carbaldehyde
SMILES O=CC1(CCOCC1)C1=CC=CC=C1

Specifications

Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 190.24
Percent Purity 95%
Chemical Name or Material 4-Phenyltetrahydropyran-4-carboxaldehyde
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only