missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylphenol, 97%

Catalog No. AC158712500
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC158712500 250 g Plastic bottle
AC158710010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC158712500 Supplier Thermo Scientific Chemicals Supplier No. 158712500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 92-69-3 | C12H10O | 170.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 92-69-3
Molecular Formula C12H10O
Molecular Weight (g/mol) 170.21
MDL Number MFCD00002347
InChI Key YXVFYQXJAXKLAK-UHFFFAOYSA-N
Synonym 4-hydroxybiphenyl, 1,1'-biphenyl-4-ol, biphenyl-4-ol, p-phenylphenol, 4-biphenylol, p-hydroxybiphenyl, 4-hydroxydiphenyl, p-hydroxydiphenyl, p-biphenylol, 4-diphenylol
PubChem CID 7103
ChEBI CHEBI:34422
IUPAC Name 4-phenylphenol
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point >163.0°C
Color White to Yellow
Boiling Point 307.0°C
Flash Point 165°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Linear Formula C6H5C6H4OH
Quantity 250 g
Beilstein 06,674
Merck Index 15,7416
Solubility Information Solubility in water: 0.7g/L (20°C). Other solubilities: soluble in alkalies,soluble in most common organic solvents
Formula Weight 170.21
Percent Purity 97%
Physical Form Flakes
Chemical Name or Material 4-Phenylphenol, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes
EINECSNumber 202-179-2
RTECSNumber DV5850000
TSCA TSCA

RUO – Research Use Only