missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Phenylbutylamine 98.0+%, TCI America™
Supplier: TCI America A11235ML
Specifications
| 4-Phenylbutylamine | |
| Yellow | |
| C10H15N | |
| 5 mL | |
| AGNFWIZBEATIAK-UHFFFAOYSA-N | |
| 4-phenylbutan-1-amine | |
| 83242 | |
| ≥98.0% (GC,T) |
| 13214-66-9 | |
| 144°C | |
| MFCD00008231 | |
| 1-Amino-4-phenylbutane | |
| C1=CC=C(C=C1)CCCCN | |
| 149.237 | |
| 149.24 | |
| Liquid |
Chemical Identifiers
| 13214-66-9 | |
| 149.237 | |
| AGNFWIZBEATIAK-UHFFFAOYSA-N | |
| 83242 | |
| C1=CC=C(C=C1)CCCCN |
| C10H15N | |
| MFCD00008231 | |
| 1-Amino-4-phenylbutane | |
| 4-phenylbutan-1-amine |
Safety and Handling
TSCA : No