missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylbutylamine 98.0+%, TCI America™

Catalog No. p-7117256
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A11235ML 5 mL
A112325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A11235ML Supplier TCI America Supplier No. A11235ML
Only null left

Chemical Identifiers

CAS 13214-66-9
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.237
MDL Number MFCD00008231
InChI Key AGNFWIZBEATIAK-UHFFFAOYSA-N
Synonym 1-Amino-4-phenylbutane
PubChem CID 83242
IUPAC Name 4-phenylbutan-1-amine
SMILES C1=CC=C(C=C1)CCCCN

Specifications

Color Yellow
Boiling Point 144°C
Quantity 5 mL
Formula Weight 149.24
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Phenylbutylamine