accessibility menu, dialog, popup

UserName

H2N

CAS RN 13214-66-9

IUPAC Name: 4-phenylbutan-1-amine
Synonyms: 4-phenylbutylamine
Molecular Weight (g/mol): 149.24
Molecular Formula: C10H15N
InChi Key: AGNFWIZBEATIAK-UHFFFAOYSA-N
SMILES: NCCCCC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results