missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylbenzophenone 98.0+%, TCI America™

Catalog No. P111625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P111625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P111625G Supplier TCI America Supplier No. P111625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2128-93-0
Molecular Formula C19H14O
Molecular Weight (g/mol) 258.32
MDL Number MFCD00003079
InChI Key LYXOWKPVTCPORE-UHFFFAOYSA-N
Synonym 4-benzoylbiphenyl, 4-phenylbenzophenone, p-phenylbenzophenone, benzophenone, 4-phenyl, p-benzoylbiphenyl, 4-diphenyl phenyl ketone, p-biphenylyl phenyl ketone, trigonal 12, 4-biphenylyl phenyl ketone, methanone, 1,1'-biphenyl-4-ylphenyl
PubChem CID 75040
IUPAC Name phenyl-(4-phenylphenyl)methanone
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)C3=CC=CC=C3

Specifications

Melting Point 102°C
Color White-Yellow
Boiling Point 245°C
Quantity 25 g
Formula Weight 258.32
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Phenylbenzophenone
RTECSNumber PC4936800
TSCA Yes