missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-3-butyn-2-one, 96%

Catalog No. AAH2782603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2782603 1 g
AAH2782606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2782603 Supplier Thermo Scientific Chemicals Supplier No. H2782603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1817-57-8 | C10H8O | 144.173 g/mol

It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1817-57-8
Molecular Formula C10H8O
Molecular Weight (g/mol) 144.173
MDL Number MFCD00008776
InChI Key UPEUQDJSUFHFQP-UHFFFAOYSA-N
Synonym 4-phenyl-3-butyn-2-one, 3-butyn-2-one, 4-phenyl, 1-phenyl-1-butyn-3-one, phenylbutynone, acetylphenyl-acetylene, acmc-1boyr, 4-phenyl-3-butyn-2-on, 4-phenyl-3-batyu-2-one, 4-phenyl-3-butyn-2-0ne, 4-phenyl-but-3-yn-2-one
PubChem CID 74555
ChEBI CHEBI:51731
IUPAC Name 4-phenylbut-3-yn-2-one
SMILES CC(=O)C#CC1=CC=CC=C1

Specifications

Density 0.99
Boiling Point 75°C to 76°C (0.8 mmHg)
Flash Point 95°C (203°F)
Refractive Index 1.574
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 144.17
Percent Purity 96%
Chemical Name or Material 4-Phenyl-3-butyn-2-one
Hazard Category H302
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 217-327-1
RTECSNumber ES0890000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only