missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-1-cyclohexene, ≥96.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014330
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014330 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014330 Supplier Merck Emd Millipore Supplier No. 0559525MG

4-Phenyl-1-cyclohexene is an analog of vinylcyclohexene.

Specifications

CAS 4994-16-5
Linear Formula C12H14
MDL Number MFCD00049188
Synonym 3-Cyclohexen-1-ylbenzene; 4-Phenylcyclohexene
Type Analytical Standard
Quantity 25 mg
Formula Weight 158.24
Percent Purity ≥96.0% (GC)
Grade Analytical Standard