missing translation for 'onlineSavingsMsg'
Learn More

4-(Phenoxymethyl)benzoic acid, 97%, Thermo Scientific™

Numéro de catalogue. CC63701CB
Modifier l'affichage
Click to view available options
Quantité:
250mg
1g
10g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CC63701CB 250mg
CC63701DA 1g
CC63701EA 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. CC63701CB Fournisseur Thermo Scientific Chemicals Code fournisseur CC63701CB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 31719-76-3
Formule moléculaire C14H11O3
Poids moléculaire (g/mol) 227.24
Numéro MDL MFCD00623268
Clé InChI GRBUVHSYBRTCIB-UHFFFAOYSA-M
Synonyme 4-phenoxymethyl benzoic acid, 4-phenoxymethyl benzenecarboxylic acid, 4-phenoxymethyl-benzoic acid, phenoxymethylbenzenecarboxylicacid
PubChem CID 737267
Nom de l’IUPAC 4-(phenoxymethyl)benzoic acid
SOURIRES [O-]C(=O)C1=CC=C(COC2=CC=CC=C2)C=C1

Spécifications

Nom ou substance chimique 4-(Phenoxymethyl)benzoic acid
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Quantité 250mg
Poids de formule 228.25
Pourcentage de pureté 97%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.