missing translation for 'onlineSavingsMsg'
Learn More

4-Phenoxybenzophenone 95.0+%, TCI America™

Numéro de catalogue. P13285G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
P13285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. P13285G Fournisseur TCI America Code fournisseur P13285G
Il en reste null

Identifiants chimiques

CAS 6317-73-3
Molecular Formula C19H14O2
Molecular Weight (g/mol) 274.319
MDL Number MFCD00017553
InChI Key ITVUPWDTDWMACZ-UHFFFAOYSA-N
Synonym 4-phenoxybenzophenone, 4-phenoxyphenyl phenyl methanone, unii-1oh6xt406c, 4-phenoxyphenyl-phenylmethanone, p-phenoxybenzophenone, 4-phenoxy benzophenone, 4-benzoyldiphenyl ether, acmc-209ndn, benzophenone, 4-phenoxy, phenyl 4-phenoxyphenyl ketone
PubChem CID 238623
IUPAC Name (4-phenoxyphenyl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)OC3=CC=CC=C3

Spécifications

Melting Point 64°C
Color Yellow
Quantity 5 g
Formula Weight 274.32
Physical Form Crystalline Powder
Chemical Name or Material 4-Phenoxybenzophenone