missing translation for 'onlineSavingsMsg'
Learn More

4-Phenoxybenzoic acid, 98%, Thermo Scientific™

Numéro de catalogue. p-3758654
Modifier l'affichage
Click to view available options
Quantité:
1g
5g
25g
Emballage:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
AC352720010 1g Glass bottle
AC352720050 5g Glass bottle
AC352720250 25g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. AC352720010 Fournisseur Thermo Scientific Chemicals Code fournisseur 352720010

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Identifiants chimiques

CAS 2215-77-2
Formule moléculaire C13H10O3
Poids moléculaire (g/mol) 214.22
Numéro MDL MFCD00002539
Clé InChI RYAQFHLUEMJOMF-UHFFFAOYSA-N
Synonyme p-phenoxybenzoic acid, benzoic acid, 4-phenoxy, diphenyl ether 4-carboxylic acid, 4-phenoxy-benzoic acid, 4-carboxybiphenyl ether, 4-carboxydiphenyl ether, benzoic acid, p-phenoxy, p-carboxydiphenyl ether, 4-phenoxy benzoic acid, 4-phoc6h4cooh
PubChem CID 75182
ChEBI CHEBI:72632
Nom de l’IUPAC 4-phenoxybenzoic acid
SOURIRES OC(=O)C1=CC=C(OC2=CC=CC=C2)C=C1

Spécifications

Nom ou substance chimique 4-Phenoxybenzoic acid
CAS % minimum 97.5
CAS % maximal 100.0
Spectre infrarouge Authentic
Fourchette de pourcentages d’essai 98%
Emballage Glass bottle
Formule linéaire C6H5OC6H4CO2H
Quantité 1g
Poids de formule 214.22
Pourcentage de pureté 98%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 218-682-5

RUO – Research Use Only