missing translation for 'onlineSavingsMsg'
Learn More

4-Pentylbenzoic acid, 97%, Thermo Scientific™

Numéro de catalogue. BTB09863EB
Modifier l'affichage
Click to view available options
Quantité:
1g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
BTB09863EB 25g
BTB09863DA 1g
BTB09863EA 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. BTB09863EB Fournisseur Thermo Scientific Chemicals Code fournisseur BTB09863EB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 26311-45-5
Formule moléculaire C12H16O2
Poids moléculaire (g/mol) 192.258
Numéro MDL MFCD00002572
Clé InChI CWYNKKGQJYAHQG-UHFFFAOYSA-N
Synonyme benzoic acid, 4-pentyl, 4-n-pentylbenzoic acid, p-pentylbenzoic acid, 4-amylbenzoic acid, p-amylbenzoic acid, para-pentylbenzoic acid, benzoic acid, p-pentyl, 4-pentyl benzoic acid, p-n-pentylbenzoic acid, 4-pentylbenzoicacid
PubChem CID 33479
Nom de l’IUPAC 4-pentylbenzoic acid
SOURIRES CCCCCC1=CC=C(C=C1)C(=O)O

Spécifications

Nom ou substance chimique 4-Pentylbenzoic acid
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber glass bottle
Formule linéaire CH3(CH2)4C6H4CO2H
Quantité 25g
Poids de formule 192.26
Pourcentage de pureté 97%
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.