missing translation for 'onlineSavingsMsg'
Learn More

4-Penten-1-ol 98.0+%, TCI America™

Catalog No. P124325ML
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P124325ML 25 mL
P1243250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P124325ML Supplier TCI America Supplier No. P124325ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 821-09-0
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.134
MDL Number MFCD00002975
InChI Key LQAVWYMTUMSFBE-UHFFFAOYSA-N
Synonym 4-penten-1-ol, 4-pentenol, 4-pentenyl alcohol, 2-allylethyl alcohol, unii-w0558tqc6f, 4-pentene-1-ol, 4-pentenol-1, 4-penten-1ol, 4penten-1-ol, 5-hydroxy-1-pentene
PubChem CID 13181
IUPAC Name pent-4-en-1-ol
SMILES C=CCCCO

Specifications

Color Colorless
Boiling Point 138°C
Quantity 25 mL
UN Number 1987
Formula Weight 86.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Penten-1-ol
EINECSNumber (2)-2393
TSCA No