missing translation for 'onlineSavingsMsg'
Learn More

4-Oxocyclohexanecarboxylic acid, 98%, Thermo Scientific Chemicals

Catalog No. AAH2729403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2729403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2729403 Supplier Thermo Scientific Chemicals Supplier No. H2729403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 874-61-3 | Molecular Formula: C7H10O3 | Molecular Weight: 142.154 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 874-61-3
Molecular Formula C7H10O3
Molecular Weight (g/mol) 142.154
MDL Number MFCD00102035
InChI Key OWLXUYGCLDGHJJ-UHFFFAOYSA-N
Synonym 4-oxocyclohexanecarboxylic acid, 4-ketocyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 4-oxo, 4-oxo-chca, cyclohexanone-4-carboxylic acid, 4-oxocyclohexane carboxylic acid, 4-oxocyclohexanecarboxylate, 4-oxocyclohexanecarboxylicacid, 4-oxo-cyclohexanecarboxylic acid, cyclohexanecarboxylicacid, 4-oxo
PubChem CID 192730
ChEBI CHEBI:1921
IUPAC Name 4-oxocyclohexane-1-carboxylic acid
SMILES C1CC(=O)CCC1C(=O)O

Specifications

Melting Point 65°C to 71°C
Quantity 1 g
Formula Weight 142.16
Percent Purity 98%
Chemical Name or Material 4-Oxocyclohexanecarboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only