missing translation for 'onlineSavingsMsg'
Learn More

4-Nitroveratrole, 98+%

Catalog No. AAA1826414
Change view
Click to view available options
Quantity:
25 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1826414 25 g
AAA1826418 50 g
AAA1826430 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1826414 Supplier Thermo Scientific Chemicals Supplier No. A1826414

CAS: 709-09-1 | C8H9NO4 | 183.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 709-09-1
Molecular Formula C8H9NO4
Molecular Weight (g/mol) 183.16
MDL Number MFCD00007238
InChI Key YFWBUVZWCBFSQN-UHFFFAOYSA-N
Synonym 3,4-dimethoxynitrobenzene, 4-nitroveratrole, benzene, 1,2-dimethoxy-4-nitro, 4-nitrcveratrole, 4-nitro-1,2-dimethoxybenzene, benzene,2-dimethoxy-4-nitro, 1,2-dimethoxy-4-nitrobenzol, 3,4-dimethoxylnitrobenzene, 3,4-dimethoxy-1-nitrobenzene
PubChem CID 69728
IUPAC Name 1,2-dimethoxy-4-nitrobenzene
SMILES COC1=CC=C(C=C1OC)[N+]([O-])=O

Specifications

Melting Point 95°C to 98°C
Boiling Point 221°C to 223°C (10 mmHg)
Quantity 25 g
Beilstein 1878614
Formula Weight 183.16
Percent Purity ≥98%
Chemical Name or Material 4-Nitroveratrole
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 211-906-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only