missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrosalicylic Acid 98.0+%, TCI America™

Numéro de catalogue. N02565G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
N02565G 5 g
N025625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. N02565G Fournisseur TCI America Code fournisseur N02565G
Il en reste null

Identifiants chimiques

CAS 619-19-2
Molecular Formula C7H5NO5
Molecular Weight (g/mol) 183.12
MDL Number MFCD00071505
InChI Key UKWUOTZGXIZAJC-UHFFFAOYSA-N
Synonym 4-nitrosalicylic acid, 2-hydroxy-4-nitrobenzoic acid, benzoic acid, 2-hydroxy-4-nitro, 4-nitro-2-hydroxybenzoic acid, p-nitrosalicylic acid, salicylic acid, 4-nitro, 4-nitro-salicylsaure, 4-nitro-salicylsaure german, 2-hydroxy-4-nitro-benzoic acid, p-nitrosalicylsaure
PubChem CID 69266
IUPAC Name 2-hydroxy-4-nitrobenzoic acid
SMILES OC(=O)C1=CC=C(C=C1O)[N+]([O-])=O

Spécifications

Melting Point 234°C
Quantity 5 g
Formula Weight 183.12
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrosalicylic Acid
RTECSNumber VO5300100
TSCA Yes